Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. D0577100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0577100G 100 g
D057725G 25 g
D0577500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0577100G Supplier TCI America Supplier No. D0577100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 611-99-4
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002358
InChI Key RXNYJUSEXLAVNQ-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy
PubChem CID 69150
ChEBI CHEBI:34365
IUPAC Name bis(4-hydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O

Specifications

Melting Point 221°C
Quantity 100 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxybenzophenone
EINECSNumber (4)-0132
RTECSNumber DJ0880000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.