Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihydroxydiphenylmethane 99.0+%, TCI America™
SDP

Catalog No. B081925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B081925G 25 g
B08195G 5 g
B0819100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B081925G Supplier TCI America Supplier No. B081925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 620-92-8
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002385
InChI Key PXKLMJQFEQBVLD-UHFFFAOYSA-N
Synonym 4,4'-methylenediphenol, bis 4-hydroxyphenyl methane, 4,4'-dihydroxydiphenylmethane, phenol, 4,4'-methylenebis, p-p-hydroxybenzyl phenol, bis p-hydroxyphenyl methane, phenol, 4,4'-methylenedi, 4,4'-methylenebisphenol, p,p'-bis hydroxyphenyl methane, 4-4-hydroxyphenyl methyl phenol
PubChem CID 12111
ChEBI CHEBI:34575
IUPAC Name 4-[(4-hydroxyphenyl)methyl]phenol
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)O)O

Specifications

Melting Point 160°C
Quantity 25 g
Formula Weight 200.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxydiphenylmethane
EINECSNumber (4)-0090
RTECSNumber SL9625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.