Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihydroxytetraphenylmethane 98.0+%, TCI America™
SDP

Catalog No. D271125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D271125G 25 g
D27115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D271125G Supplier TCI America Supplier No. D271125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1844-01-5
Molecular Formula C25H20O2
Molecular Weight (g/mol) 352.433
MDL Number MFCD00045785
InChI Key BATCUENAARTUKW-UHFFFAOYSA-N
Synonym Bis(4-hydroxyphenyl)diphenylmethane, (Diphenyl)bis(4-hydroxyphenyl)methane
PubChem CID 232881
IUPAC Name 4-[(4-hydroxyphenyl)-diphenylmethyl]phenol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 352.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxytetraphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.