Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diiodobiphenyl 98.0+%, TCI America™
SDP

Catalog No. D2315250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2315250G 250 g
D231525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2315250G Supplier TCI America Supplier No. D2315250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3001-15-8
Molecular Formula C12H8I2
Molecular Weight (g/mol) 406.005
MDL Number MFCD00001057
InChI Key GPYDMVZCPRONLW-UHFFFAOYSA-N
Synonym 4,4'-diiodobiphenyl, 4,4'-diiodo-1,1'-biphenyl, 4,4'-diiododiphenyl, 1,1'-biphenyl, 4,4'-diiodo, 1-iodo-4-4-iodophenyl benzene, 4,4-diiodobiphenyl, 4-4'-iodophenyl iodobenzene, 4,4;-diiodobiphenyl, 4,4'-diiododophenyl, pract, 4-iodo-1-4-iodophenyl benzene
PubChem CID 76348
IUPAC Name 1-iodo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)I

Specifications

Melting Point 208°C
Color Yellow
Quantity 250 g
UN Number 3152
Formula Weight 406.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diiodobiphenyl
EINECSNumber (4)-1507
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.