Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxybenzhydrol 98.0+%, TCI America™
SDP

Catalog No. D214125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D214125G 25 g
D21415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D214125G Supplier TCI America Supplier No. D214125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 728-87-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008410
InChI Key ZODAOVNETBTTJX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh
PubChem CID 69768
IUPAC Name bis(4-methoxyphenyl)methanol
SMILES COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1

Specifications

Melting Point 70°C
Color White
Quantity 25 g
Formula Weight 244.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxybenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.