Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxybenzhydrylamine 98.0+%, TCI America™
SDP

Catalog No. D220225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D220225G 25 g
D22021G 1 g
D22025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D220225G Supplier TCI America Supplier No. D220225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19293-62-0
Molecular Formula C15H17NO2
Molecular Weight (g/mol) 243.31
MDL Number MFCD00191409
InChI Key HROGQYMZWGPHIB-UHFFFAOYSA-N
Synonym 1,1-Di(p-anisyl)methylamine
PubChem CID 350042
IUPAC Name 1,1-bis(4-methoxyphenyl)methanamine
SMILES COC1=CC=C(C=C1)C(N)C1=CC=C(OC)C=C1

Specifications

Melting Point 62°C
Color White
Quantity 25 g
Formula Weight 243.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxybenzhydrylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.