Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethoxybutyronitrile, 98%, Thermo Scientific™

Catalog No. AAB2351206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2351206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2351206 Supplier Thermo Scientific Chemicals Supplier No. B2351206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14618-78-1
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00019883
InChI Key XJHNCGVHPLDMRK-UHFFFAOYSA-N
Synonym 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f
PubChem CID 84558
IUPAC Name 4,4-dimethoxybutanenitrile
SMILES COC(CCC#N)OC

Specifications

Boiling Point 90°C to 91°C (14 mmHg)
Quantity 5 g
UN Number UN3276
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material 4,4-Dimethoxybutyronitrile
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.