Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxytrityl Chloride, 98%

Catalog No. p-3736271
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146020050 5 g Glass bottle
AC146020250 25 g Glass bottle
AC146021000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC146020050 Supplier Thermo Scientific Chemicals Supplier No. 146020050
Only null left
Add to Cart

CAS: 40615-36-9 | C21H19ClO2 | 338.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40615-36-9
Molecular Formula C21H19ClO2
Molecular Weight (g/mol) 338.83
MDL Number MFCD00008409
InChI Key JBWYRBLDOOOJEU-UHFFFAOYSA-N
Synonym 4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
PubChem CID 96831
IUPAC Name 1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121°C to 127°C
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C(C6H4OCH3)2Cl
Quantity 5 g
Fieser 01,188
Formula Weight 338.82
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dimethoxytrityl chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 255-002-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.