Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, trisodium salt, 98%, pure, anhydrous

Catalog No. AC391975000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00012462
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Synonym sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous
PubChem CID 6224
ChEBI CHEBI:53258
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point 300.0°C
Color White
Flash Point 290°C
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic bottle
Quantity 500 g
Solubility Information Solubility in water: 500g/L (20°C).
Formula Weight 258.07
Percent Purity 0.98
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Citric acid, trisodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.