Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethyl-2,2'-bipyridyl 1-Oxide 98.0+%, TCI America™
SDP

Catalog No. D44671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44671G 1 g
D44675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44671G Supplier TCI America Supplier No. D44671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81998-03-0
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD11042487
InChI Key VNSKKRAUVGCMPH-UHFFFAOYSA-N
PubChem CID 13586792
IUPAC Name 4-methyl-2-(4-methylpyridin-2-yl)-1-oxidopyridin-1-ium
SMILES CC1=CC(=NC=C1)C2=[N+](C=CC(=C2)C)[O-]

Specifications

Melting Point 87°C
Color Yellow
Quantity 1 g
Synonym 4,4′C-Dimethyl-2,2′C-bipyridine 1-Oxide
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethyl-2,2′-bipyridyl 1-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.