Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-2-cyclohexen-1-one 96.0+%, TCI America™
SDP

Catalog No. D385225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D385225G 25 g
D38525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D385225G Supplier TCI America Supplier No. D385225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1073-13-8
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00009695
InChI Key HAUNPYVLVAIUOO-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
PubChem CID 136839
IUPAC Name 4,4-dimethylcyclohex-2-en-1-one
SMILES CC1(C)CCC(=O)C=C1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 25 g
Formula Weight 124.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-2-cyclohexen-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.