Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4-Dimethyl-2-pentanone

Catalog No. 13687525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-687-525G 25 g
13-687-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-687-525G Supplier Sigma Aldrich Supplier No. 13687525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4,4-dimethyl-2-pentanone was used as starting reagent in the synthesis of 3,4,4,5-tetramethyl-3-hexanol
  • It was used in the gas-phase reactions of oh radicals with 2,2,4-trimethylpentane

Specifications

CAS 590-50-1
Density 0.809 g/mL (at 25°C (literature))
Boiling Point 125°C to 130°C (lit.)
Molecular Formula C7H14O
Linear Formula (CH3)3 CCH2COCH3
Refractive Index n20/D 1.404 (literature)
Quantity 25 g
MDL Number MFCD00008854
Synonym Methyl neopentyl ketone
Molecular Weight (g/mol) 114.19
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.