Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-2-pentanone, 99%

Catalog No. AC159710250
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AC159710250

Supplier: Thermo Scientific Chemicals 159710250

Only null left
Add to Cart

CAS: 590-50-1 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-50-1
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
InChI Key AZASWMGVGQEVCS-UHFFFAOYSA-N
PubChem CID 11546
IUPAC Name 4,4-dimethylpentan-2-one
SMILES CC(=O)CC(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8040g/mL
Boiling Point 125.0°C to 130.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4025 to 1.4045
Linear Formula (CH3)3CCH2COCH3
Quantity 25 g
Beilstein 01,702
Specific Gravity 0.804
Synonym 4,4-dimethyl-2-pentanone, 2-pentanone, 4,4-dimethyl, methyl neopentyl ketone, neopentyl methyl ketone, methyl neoamyl ketone, 4,4-dimethyl-pentan-2-one, 2-pentanone,4,4-dimethyl, acmc-1awgm, neo-c5h11coch3
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4, 4-Dimethyl-2-pentanone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 209-685-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.