Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethylbenzophenone, 98+%

Catalog No. AAA1411114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1411114 25 g
AAA1411106 5 g
AAA1411122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1411114 Supplier Thermo Scientific Chemicals Supplier No. A1411114
Add to Cart

CAS: 611-97-2 | C15H14O | 210.276 g/mol

4,4'-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-97-2
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017214
InChI Key ZWPWLKXZYNXATK-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzophenone, p-tolyl ketone, bis 4-methylphenyl methanone, di-p-tolyl ketone, methanone, bis 4-methylphenyl, p,p'-dimethylbenzophenone, p,p'-dimethyl di-phenyl ketone, bis-p-tolylmethanone, benzophenone, 4,4'-dimethyl, 4,4-dimethylbenzophenone
PubChem CID 69148
IUPAC Name bis(4-methylphenyl)methanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 92°C to 96°C
Boiling Point 200°C (17 mmHg)
Quantity 25 g
Beilstein 1240527
Solubility Information Insoluble in water.
Formula Weight 210.28
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dimethylbenzophenone
EINECSNumber 210-287-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.