Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dithiodianiline 98.0+%, TCI America™
SDP

Catalog No. D029125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D029125G 25 g
D02915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D029125G Supplier TCI America Supplier No. D029125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 722-27-0
Molecular Formula C12H12N2S2
Molecular Weight (g/mol) 248.362
MDL Number MFCD00007882
InChI Key MERLDGDYUMSLAY-UHFFFAOYSA-N
Synonym 4,4'-dithiodianiline, 4-aminophenyl disulfide, benzenamine, 4,4'-dithiobis, 4,4'-dithiobisaniline, 4,4'-diaminodiphenyl disulfide, p,p'-dithiodianiline, bis p-aminophenyl disulfide, bis 4-aminophenyl disulfide, p,p'-dithiobisaniline, 4-4-aminophenyl disulfanyl aniline
PubChem CID 12875
IUPAC Name 4-[(4-aminophenyl)disulfanyl]aniline
SMILES C1=CC(=CC=C1N)SSC2=CC=C(C=C2)N

Specifications

Melting Point 75°C
Quantity 25 g
Formula Weight 248.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dithiodianiline
EINECSNumber (3)-1127
RTECSNumber BX9580000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.