Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 98.0+%, TCI America™
SDP

Catalog No. H1314100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1314100G 100 g
H131425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1314100G Supplier TCI America Supplier No. H1314100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-48-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00048232
InChI Key OJYLAHXKWMRDGS-UHFFFAOYSA-N
Synonym zingerone, vanillylacetone, zingiberone, 4-4-hydroxy-3-methoxyphenyl butan-2-one, zingherone, 4-4-hydroxy-3-methoxyphenyl-2-butanone, gingerone, vanillyl acetone, 0-paradol, 2-butanone, 4-4-hydroxy-3-methoxyphenyl
PubChem CID 31211
ChEBI CHEBI:68657
IUPAC Name 4-(4-hydroxy-3-methoxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 148°C
Quantity 100 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxy-3-methoxyphenyl)-2-butanone
RTECSNumber EL8900000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.