Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Hydroxybutyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. H114225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H114225G 25 g
H11425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H114225G Supplier TCI America Supplier No. H114225G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 59801-41-1
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.29
MDL Number MFCD09038525
InChI Key STKAHXPYHDVHGO-UHFFFAOYSA-N
Synonym 4-(1,1-Dioxothiomorpholino)-1-butanol
PubChem CID 12324072
IUPAC Name 4-(4-hydroxybutyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES OCCCCN1CCS(=O)(=O)CC1

Specifications

Melting Point 65°C
Quantity 25 g
Formula Weight 207.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxybutyl)thiomorpholine 1,1-Dioxide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.