Learn More
4-(4-Hydroxyphenyl)-2-butanone, 98%
Description
4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 80°C to 84°C |
| Quantity | 100 g |
| Beilstein | 776080 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 164.2 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-(4-Hydroxyphenyl)-2-butanone |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 226-806-4 |
| RTECSNumber | EL8925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.