Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Hydroxyphenyl)cyclohexanone 95.0+%, TCI America™
SDP

Catalog No. H131625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H131625G 25 g
H13165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H131625G Supplier TCI America Supplier No. H131625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105640-07-1
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00210693
InChI Key SLJYPZJZQIHNGU-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl cyclohexanone, 4-4-hydroxyphenyl cyclohexan-1-one, 4-4-hydroxyphenyl-cyclohexanone, cyclohexanone, 4-4-hydroxyphenyl, 4-4-hydroxyphenyl-1-cyclohexanone, acmc-1cukv, 4-4-oxocyclohexyl phenol, ksc495c5l, 4-4'-hydroxyphenyl-cyclohexanone
PubChem CID 4598916
IUPAC Name 4-(4-hydroxyphenyl)cyclohexan-1-one
SMILES OC1=CC=C(C=C1)C1CCC(=O)CC1

Specifications

Melting Point 169°C
Color White
Quantity 25 g
Formula Weight 190.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxyphenyl)cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.