Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(4-Iodophenylsulfamoyl)acetanilide, 97%, Thermo Scientific™

Numéro de catalogue. AAH6015703
Modifier l'affichage
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH6015703 1 g
AAH60157MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH6015703 Fournisseur Thermo Scientific Chemicals Code fournisseur H6015703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 19837-94-6
Molecular Formula C14H13IN2O3S
Molecular Weight (g/mol) 416.23
MDL Number MFCD00584900
InChI Key VCYWXXUBOZNHBJ-UHFFFAOYSA-N
Synonym n-4-4-iodophenyl sulfamoyl phenyl acetamide, n-4-4-iodophenyl amino sulfonyl phenyl acetamide, cbmicro_016050, cambridge id 5261047, 4'-4-iodophenylsulfamoyl acetanilide, n-4-4-iodo-phenylsulfamoyl-phenyl-acetamide
PubChem CID 1097308
IUPAC Name N-[4-[(4-iodophenyl)sulfamoyl]phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=C(I)C=C1

Spécifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 416.23
Percent Purity 97%
Chemical Name or Material 4'-(4-Iodophenylsulfamoyl)acetanilide
TSCA No
Recommended Storage Ambient temperatures
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.