Learn More
Sigma Organic Chemistry 4- 4-Morpholinyl -1 3 5 | 1G | 2045-25-2 | MFCD00023307
Supplier: Sigma Organic Chemistry CBR001781G

4- 4-Morpholinyl -1 3 5 , 1G
About this Item:
CAS #: 2045-25-2
MDL #: MFCD00023307
Molecular Weight: 181.2
UNSPSC Code: 12352200
Chemical Formula: C7H11N5O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.