Learn More
4-(4-Nitrobenzyl)pyridine, 98+%
Description
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Specifications
| Melting Point | 69°C to 72°C |
| Quantity | 25 g |
| Beilstein | 170877 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in acetone. Insoluble in water. |
| Formula Weight | 214.22 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-(4-Nitrobenzyl)pyridine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335-H340 |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c |
| EINECSNumber | 214-108-2 |
| RTECSNumber | UT6360000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.