Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-Oxydianiline

Catalog No. 501839170
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501839170 100 g
50-290-3957 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501839170 Supplier Sigma Aldrich Supplier No. 516805100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Oxydianiline
TRUSTED_SUSTAINABILITY

Specifications

CAS 101-80-4
Melting Point 188°C to 192°C (lit.)
Molecular Formula C12H12N2O
Linear Formula O(C6H4NH2)2
MDL Number MFCD00007863
Quantity 100 g
Synonym 4,4 -Diaminodiphenyl ether; 4-Aminophenyl ether
Molecular Weight (g/mol) 200.24
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.