Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Phenyl-1H-pyrazol-1-yl)piperidine, 95%, Thermo Scientific™

Catalog No. AAH5080503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5080503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5080503 Supplier Thermo Scientific Chemicals Supplier No. H5080503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(4-Phenyl-1H-pyrazol-1-yl)piperidine is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-40-2
Molecular Formula C14H17N3
Molecular Weight (g/mol) 227.311
MDL Number MFCD08060998
InChI Key SVKHSGABZPOEQY-UHFFFAOYSA-N
Synonym 4-4-phenyl-1h-pyrazol-1-yl piperidine, 4-4-phenylpyrazol-1-yl piperidine, 4-phenyl-1-4-piperidyl pyrazole, 4-4-phenyl-pyrazol-1-yl-piperidine
PubChem CID 24208827
IUPAC Name 4-(4-phenylpyrazol-1-yl)piperidine
SMILES C1CNCCC1N2C=C(C=N2)C3=CC=CC=C3

Specifications

Quantity 1 g
Formula Weight 227.31
Percent Purity 95%
Chemical Name or Material 4-(4-Phenyl-1H-pyrazol-1-yl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.