Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Piperidinyloxy)benzonitrile, 98%

Catalog No. p-7033789
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50958MD 250 mg
AAH5095803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50958MD Supplier Thermo Scientific Chemicals Supplier No. H50958MD
Only null left
Add to Cart

CAS: 224178-67-0 | C12H14N2O | 202.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 224178-67-0
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD07368391
InChI Key DRIREFRFHFBPIN-UHFFFAOYSA-N
PubChem CID 11241109
IUPAC Name 4-piperidin-4-yloxybenzonitrile
SMILES C1CNCCC1OC2=CC=C(C=C2)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Synonym 4-piperidin-4-yloxy benzonitrile, 4-4-piperidinyloxy benzonitrile, 4-4-piperidyloxy benzonitrile, 4-piperidin-4-yloxy-benzonitrile, acmc-1cb1d, benzonitrile,4-4-piperidinyloxy, benzonitrile, 4-4-piperidinyloxy, 4-piperidin-4-yl-oxy-benzonitrile
Formula Weight 202.26
Percent Purity 98%
Chemical Name or Material 4-(4-Piperidinyloxy)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.