Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Sulfonylbis-(methylbenzoate) (Tech.), 90%, Thermo Scientific™

Catalog No. AC123492500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC123492500 250 g
AC123490500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC123492500 Supplier Thermo Scientific Chemicals Supplier No. 123492500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3965-53-5
Molecular Formula C16H14O6S
Molecular Weight (g/mol) 334.342
InChI Key YQYRQRUZQNRJIW-UHFFFAOYSA-N
Synonym dimethyl 4,4'-sulphonyldibenzoate, dimethyl 4,4'-sulfonyldibenzoate, benzoic acid, 4,4'-sulfonylbis-, dimethyl ester, benzoic acid, 4,4'-sulfonyldi-, dimethyl ester, methyl 4-4-methoxycarbonyl phenyl sulfonyl benzoate, 4,4'-sulfonylbis methyl benzoate, 4,4'-sulfonylbis benzoic acid methyl ester, benzoic acid, sulfonyl di-p,p'-dimethyl ester, methyl 4-4-methoxycarbonylphenyl sulfonylbenzoate, benzoic acid,4,4'-sulfonylbis-, 1,1'-dimethyl ester
PubChem CID 77583
IUPAC Name methyl 4-(4-methoxycarbonylphenyl)sulfonylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(C=C2)C(=O)OC

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 193°C to 200°C
Color Gray to White
Infrared Spectrum Authentic
Linear Formula (CH3OOC6H4)2SO2
Quantity 250 g
Beilstein 10, II, 109
Formula Weight 334.33
Percent Purity ≥88%
Physical Form Powder
Chemical Name or Material 4, 4'-Sulfonylbis-(methylbenzoate), tech., 90%
Show More Show Less
EINECSNumber 223-577-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.