Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega 4/40 Wash Solution for Wizard MagneSil Tfx™ System

Catalog No. PRA2221 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
115 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2221 115 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2221 Supplier Promega Supplier No. A2221
Add to Cart
Add to Cart

Buffer A (BWA) is a component of the Maxwell HT DNA FFPE Isolation System (Cat.# A6372) available as a standalone item.

  • Reagent Used in DNA Purification Protocols

Specifications

Content And Storage 15°C to 30°C
Quantity 115 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.