Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]morpholine, Thermo Scientific™

Catalog No. CC17539DA
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC17539DA 1 g Amber Glass Bottle
CC17539DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC17539DA Supplier Thermo Scientific Chemicals Supplier No. CC17539DA
Add to Cart

Chemical Identifiers

CAS 485799-04-0
Molecular Formula C15H23BN2O3
Molecular Weight (g/mol) 290.17
MDL Number MFCD04039874
InChI Key ZGDLVKWIZHHWIR-UHFFFAOYSA-N
Synonym 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 6-morpholin-4-yl pyridine-3-boronic acid pinacol ester, 6-morpholinopyridin-3-ylboronic acid pinacol ester, 2-morpholinopyridine-5-boronic acid, pinacol ester, 4-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-morpholine, 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl morpholine, 2-4-morpholino pyridine-5-boronic acid pinacol ester, 4-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 6-morpholinopyridine-3-boronic acid pinacol ester
PubChem CID 2795363
IUPAC Name 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 290.17
Percent Purity 97%
Chemical Name or Material 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.