Learn More
Sigma Organic Chemistry 4-5-4-fluorophenyl -1 3 | 100mg | MFCD03424488
Supplier: Sigma Organic Chemistry CDS010866100MG

4-5-4-fluorophenyl -1 3 , 100mg
About this Item:
MDL #: MFCD03424488
Molecular Weight: 247.27
UNSPSC Code: 12352200
Chemical Formula: C13H14FN3O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.