Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,6,7-Tetrabromophenolphthalein 80.0+%, TCI America™
SDP

Catalog No. T00435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00435G 5 g
T00431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00435G Supplier TCI America Supplier No. T00435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13027-28-6
Molecular Formula C20H10Br4O4
Molecular Weight (g/mol) 633.91
MDL Number MFCD00047314
InChI Key GNJLVMKERYSIAI-UHFFFAOYSA-N
PubChem CID 83058
IUPAC Name 4,5,6,7-tetrabromo-3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C=C1

Specifications

Quantity 5 g
Formula Weight 633.91
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6,7-Tetrabromophenolphthalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.