Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine 98.0+%, TCI America™
SDP

Catalog No. T268625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T268625G 25 g
T26865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T268625G Supplier TCI America Supplier No. T268625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54903-50-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD03839944
InChI Key OGUWOLDNYOTRBO-UHFFFAOYSA-N
PubChem CID 3085076
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridine
SMILES C1CNCC2=C1SC=C2

Specifications

Color Yellow
Boiling Point 75°C
Quantity 25 g
Formula Weight 139.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.