Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane 98.0+%, TCI America™
SDP

Catalog No. O04551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04551G 1 g
O04555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04551G Supplier TCI America Supplier No. O04551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1785-64-4
Molecular Formula C16H8F8
Molecular Weight (g/mol) 352.227
MDL Number MFCD13195584
InChI Key GUHKMHMGKKRFDT-UHFFFAOYSA-N
Synonym 5,6,11,12,13,14,15,16-Octafluorotricyclo[8.2.2.2(4,7)]hexadeca-4,6,10,12,13,15-hexaene
PubChem CID 11739627
SMILES C1CC2=C(C(=C(CCC3=C(C(=C1C(=C3F)F)F)F)C(=C2F)F)F)F

Specifications

Color White
Quantity 1 g
Formula Weight 352.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.