Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene 98.0+%, TCI America™
SDP

Catalog No. B270925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B270925G 25 g
B27091G 1 g
B27095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B270925G Supplier TCI America Supplier No. B270925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 161265-03-8
Molecular Formula C39H32OP2
Molecular Weight (g/mol) 578.63
MDL Number MFCD00233866
InChI Key CXNIUSPIQKWYAI-UHFFFAOYSA-N
Synonym xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene
PubChem CID 636044
IUPAC Name [5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane
SMILES CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 229°C
Color White-Yellow
Quantity 25 g
Formula Weight 578.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.