Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Bis(hydroxymethyl)-2-phenylimidazole 95.0+%, TCI America™
SDP

Catalog No. B49321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49321G 1 g
B49325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49321G Supplier TCI America Supplier No. B49321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61698-32-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.229
MDL Number MFCD00047017
InChI Key UUQQGGWZVKUCBD-UHFFFAOYSA-N
PubChem CID 109115
IUPAC Name [4-(hydroxymethyl)-2-phenyl-1H-imidazol-5-yl]methanol
SMILES C1=CC=C(C=C1)C2=NC(=C(N2)CO)CO

Specifications

Melting Point 288°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.23
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Bis(hydroxymethyl)-2-phenylimidazole
EINECSNumber (5)-5628
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.