Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Diazafluoren-9-one, 98%

Catalog No. p-7037317
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5602803 1 g
AAH5602806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5602803 Supplier Thermo Scientific Chemicals Supplier No. H5602803
Only null left
Add to Cart

CAS: 50890-67-0 | C11H6N2O | 182.182 g/mol

4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].

Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.

Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50890-67-0
Molecular Formula C11H6N2O
Molecular Weight (g/mol) 182.182
MDL Number MFCD00046892
InChI Key PFMTUGNLBQSHQC-UHFFFAOYSA-N
Synonym 4,5-diazafluoren-9-one, 5h-cyclopenta 1,2-b:5,4-b' dipyridin-5-one, 5h-cyclopenta 2,1-b:3,4-b' dipyridin-5-one, 5h-pyrido 3',2':4,5 cyclopenta 1,2-b pyridin-5-one, pyridino 3',2'-1,2 cyclopenta 3,4-b pyridin-5-one, dafone, dafo, 4,5-diazafluorenone-0, 4,5-diaza-9h-fluoren-9-one
PubChem CID 342157
SMILES C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1

Specifications

Melting Point 214°C to 217°C
Quantity 1 g
Solubility Information Soluble in dichloromethane,tetrahydrofuran,chloroform,benzene,toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 4,5-Diazafluoren-9-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.