Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dibromothiophene-2-sulfonyl chloride, 97%

Catalog No. aaa1338403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1338403 1 g
AAA1338406 5 g
AAA1338414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1338403 Supplier Thermo Scientific Chemicals Supplier No. A1338403
Only null left
Add to Cart

CAS: 81606-31-7 | C4HBr2ClO2S2 | 340.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81606-31-7
Molecular Formula C4HBr2ClO2S2
Molecular Weight (g/mol) 340.43
MDL Number MFCD00041439
InChI Key WJYGHWXWQSCONR-UHFFFAOYSA-N
Synonym 4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride
PubChem CID 522755
IUPAC Name 4,5-dibromothiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC(Br)=C(Br)S1

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
UN Number UN3261
Beilstein 162960
Sensitivity Moisture sensitive
Formula Weight 340.45
Percent Purity 97%
Chemical Name or Material 4,5-Dibromothiophene-2-sulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.