Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dichloro-2-nitrotoluene 98.0+%, TCI America™
SDP

Catalog No. D327125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D327125G 25 g
D32715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D327125G Supplier TCI America Supplier No. D327125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7494-45-3
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.022
InChI Key AOTWCYSQDSKAQT-UHFFFAOYSA-N
PubChem CID 343795
IUPAC Name 1,2-dichloro-4-methyl-5-nitrobenzene
SMILES CC1=CC(=C(C=C1[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 62°C
Quantity 25 g
Formula Weight 206.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloro-2-nitrotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.