Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dicyanoimidazole 98.0+%, TCI America™
SDP

Catalog No. D2026250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2026250G 250 g
D202625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2026250G Supplier TCI America Supplier No. D2026250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1122-28-7
Molecular Formula C5H2N4
Molecular Weight (g/mol) 118.10
MDL Number MFCD00005194
InChI Key XGDRLCRGKUCBQL-UHFFFAOYSA-N
PubChem CID 70729
IUPAC Name 1H-imidazole-4,5-dicarbonitrile
SMILES N#CC1=C(N=CN1)C#N

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 250 g
UN Number 3439
Synonym 4,5-dicyanoimidazole, imidazole-4,5-dicarbonitrile, 4,5-imidazoledicarbonitrile, activator, pubchem7602, 4, 5-dicyanoimidazole, moa 7, imidazole-4,5-dinitrile, acmc-1brr7, 4,5-dicyano-1h-imidazole
Formula Weight 118.10
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dicyanoimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.