Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, Thermo Scientific Chemicals

Catalog No. AC378102500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC378102500 250 g
AC378100010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378102500 Supplier Thermo Scientific Chemicals Supplier No. 378102500
  • $258.00
    $209.65 / Each of 1

    Save $48.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5808-22-0 | C10H10Na2O10S2 | 400.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5808-22-0
Molecular Formula C10H10Na2O10S2
Molecular Weight (g/mol) 400.28
MDL Number MFCD00150612
InChI Key QUEAKWJKJBFNEG-UHFFFAOYSA-L
Synonym 1,8-Dihydroxynaphthalene-3,6-disulfonic acid, disodium salt dihydrate; Chromotropic acid, disodium salt dihydrate
IUPAC Name 2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
SMILES O.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]

Specifications

Alpha Vector DIHYDROXYNAPHTHALENEDISULFONICACID 021
Melting Point >300°C
Color Brown to White
Infrared Spectrum Conforms
Assay Percent Range ≥88 % (Titration acid)
Packaging glass bottle
Quantity 250 g
Solubility Information soluble aq. soln.
Formula Weight 400.28
Physical Form Crystals or Powder
Chemical Name or Material 4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.