accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHSOOOSOOOH2OH2ONaNa

CAS RN 5808-22-0

IUPAC Name: 2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
Synonyms: 2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate|Disodium Chromotropate Dihydrate
Molecular Weight (g/mol): 400.28
Molecular Formula: C10H10Na2O10S2
InChi Key: QUEAKWJKJBFNEG-UHFFFAOYSA-L
SMILES: O.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]
Molecular Weight (g/mol) 
  • (15)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (5)
  • (7)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (5)
  • (2)
  • (9)
  • (1)
  • (1)
Grade 
  • (6)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (11)
  • (15)
  • (3)
  • (2)
  • (1)
  • (1)
  • (5)
  • (7)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (9)
  • (1)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)