Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethoxy-1-benzocyclobutenecarbonitrile 98.0+%, TCI America™
SDP

Catalog No. D44855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44855G 5 g
D44851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44855G Supplier TCI America Supplier No. D44855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35202-54-1
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD01846132
InChI Key HJTHVTHXHHFXMJ-UHFFFAOYSA-N
Synonym 1-Cyano-4,5-dimethoxybenzocyclobutene, 3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile
PubChem CID 290544
IUPAC Name 3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile
SMILES COC1=C(C=C2C(CC2=C1)C#N)OC

Specifications

Melting Point 86°C
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dimethoxy-1-benzocyclobutenecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.