Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Diphenylimidazole 98.0+%, TCI America™
SDP

Catalog No. D210725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D210725G 25 g
D21075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D210725G Supplier TCI America Supplier No. D210725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 668-94-0
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00005198
InChI Key CPHGOBGXZQKCKI-UHFFFAOYSA-N
Synonym 4,5-diphenylimidazole, diphenylimidazole, 1h-imidazole, 4,5-diphenyl, imidazole, 4,5-diphenyl, dsstox_cid_25700, dsstox_rid_81072, dsstox_gsid_45700, 4,5-diphenyl-1h-imidazol, imidazole,5-diphenyl, 4,5-diphenylglyoxaline
PubChem CID 69588
IUPAC Name 4,5-diphenyl-1H-imidazole
SMILES N1C=NC(=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 234°C
Color White
Quantity 25 g
Formula Weight 220.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Diphenylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.