Learn More
4,5-Methylenedioxy-1,2-Phenylenediamine Dihydrochloride, Sigma-Aldrich
Catalog No. 01258815 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-815 100 mg
01-258-946 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-815 Supplier MilliporeSigma Supelco Supplier No. D4784100MG
Only null left
Add to Cart
Add to Cart
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride is a fluorescence tag for Alpha keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.
TRUSTED_SUSTAINABILITY
Specifications
Reagent Type Ion Pairing Reagent
Form Powder
Quantity 100 mg
Type Silylation