Learn More
4,5-Methylenedioxy-1,2-Phenylenediamine Dihydrochloride, Sigma-Aldrich
Shop All MilliporeSigma Supelco ProductsDescription
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride is a fluorescence tag for Alpha keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.
Specifications
Specifications
| Form | Powder |
| Reagent Type | Ion Pairing Reagent |
| Quantity | 50 mg |
| Type | Silylation |