Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4',6,7-Trimethoxyisoflavone 97.0+%, TCI America™
SDP

Catalog No. T34101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34101G 1 g
T34105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34101G Supplier TCI America Supplier No. T34101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 798-61-8
Molecular Formula C18H16O5
Molecular Weight (g/mol) 312.321
MDL Number MFCD00016949
InChI Key YHXIOAVHEXKZCQ-UHFFFAOYSA-N
Synonym 6,7-Dimethoxy-3-(4-methoxyphenyl)chromone
PubChem CID 688655
IUPAC Name 6,7-dimethoxy-3-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C2=COC3=CC(=C(C=C3C2=O)OC)OC

Specifications

Melting Point 179°C
Quantity 1 g
Formula Weight 312.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′,6,7-Trimethoxyisoflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.