Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4',6-Diamidino-2-phenylindole dihydrochloride, >98%, MP Biomedicals™

Catalog No. ICN15757410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15757410 10 mg
ICN15757405 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15757410 Supplier MP Biomedicals, Inc Supplier No. 0215757410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16H17Cl2N5
Molecular Weight (g/mol) 350.25
MDL Number MFCD00012681
InChI Key FPNZBYLXNYPRLR-UHFFFAOYSA-N
Synonym 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836
PubChem CID 160166
IUPAC Name dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N

Specifications

Color Yellow to Orange
Assay Percent Range >98%
Quantity 10 mg
Formula Weight 350.2
Physical Form Crystalline Powder
Chemical Name or Material 4', 6-Diamidino-2-phenylindole dihydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.