CAS RN 28718-90-3
4',6-Diamidino-2-phenylindole dihydrochloride, >98%, MP Biomedicals™
CAS: 28718-90-3 Molecular Formula: C16H17Cl2N5 Molecular Weight (g/mol): 350.25 MDL Number: MFCD00012681 InChI Key: FPNZBYLXNYPRLR-UHFFFAOYSA-N Synonym: 4',6-diamidino-2-phenylindole dihydrochloride,dapi dihydrochloride,dapi, dihydrochloride,2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride,dapi hydrochloride,2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate,2-4-amidinophenyl-6-indolecarbamidine dihydrochloride,4′,6-diamidino-2-phenylindole, 2hcl,ccris 8836 PubChem CID: 160166 IUPAC Name: dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N
MilliporeSigma™ DAPI, Dihydrochloride, Calbiochem™,
CAS: 28718-90-3 Molecular Formula: C16H17Cl2N5 Molecular Weight (g/mol): 350.25 MDL Number: MFCD00012681 InChI Key: FPNZBYLXNYPRLR-UHFFFAOYSA-N Synonym: 4',6-diamidino-2-phenylindole dihydrochloride,dapi dihydrochloride,dapi, dihydrochloride,2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride,dapi hydrochloride,2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate,2-4-amidinophenyl-6-indolecarbamidine dihydrochloride,4′,6-diamidino-2-phenylindole, 2hcl,ccris 8836 PubChem CID: 160166 IUPAC Name: dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
CAS: 28718-90-3 Molecular Formula: C16H17Cl2N5 Molecular Weight (g/mol): 350.25 MDL Number: MFCD00012681 InChI Key: FPNZBYLXNYPRLR-UHFFFAOYSA-N Synonym: 4',6-diamidino-2-phenylindole dihydrochloride,dapi dihydrochloride,dapi, dihydrochloride,2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride,dapi hydrochloride,2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate,2-4-amidinophenyl-6-indolecarbamidine dihydrochloride,4′,6-diamidino-2-phenylindole, 2hcl,ccris 8836 PubChem CID: 160166 SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N
4',6-Diamidino-2-Phenylindole Dihydrochloride MP Biomedicals
CAS: 28718-90-3 Molecular Formula: C16H17Cl2N5 Molecular Weight (g/mol): 350.25 MDL Number: MFCD00012681 InChI Key: FPNZBYLXNYPRLR-UHFFFAOYSA-N Synonym: 4',6-diamidino-2-phenylindole dihydrochloride,dapi dihydrochloride,dapi, dihydrochloride,2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride,dapi hydrochloride,2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate,2-4-amidinophenyl-6-indolecarbamidine dihydrochloride,4′,6-diamidino-2-phenylindole, 2hcl,ccris 8836 PubChem CID: 160166 IUPAC Name: dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N
2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride 90.0+%, TCI America™
CAS: 28718-90-3 Molecular Formula: C16H17Cl2N5 Molecular Weight (g/mol): 350.25 MDL Number: MFCD00012681 InChI Key: FPNZBYLXNYPRLR-UHFFFAOYSA-N Synonym: 4',6-diamidino-2-phenylindole dihydrochloride,dapi dihydrochloride,dapi, dihydrochloride,2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride,dapi hydrochloride,2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate,2-4-amidinophenyl-6-indolecarbamidine dihydrochloride,4′,6-diamidino-2-phenylindole, 2hcl,ccris 8836 PubChem CID: 160166 IUPAC Name: dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N
R&D Systems™ DAPI
A Tocris product, part of R&D Systems. Fluorescent DNA stain