Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dibromodibenzofuran 98.0+%, TCI America™
SDP

Catalog No. D48581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48581G 1 g
D4858200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48581G Supplier TCI America Supplier No. D48581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 201138-91-2
Molecular Formula C12H6Br2O
Molecular Weight (g/mol) 325.987
MDL Number MFCD00185686
InChI Key XSJLDNSNUICSQC-UHFFFAOYSA-N
PubChem CID 526362
IUPAC Name 4,6-dibromodibenzofuran
SMILES C1=CC2=C(C(=C1)Br)OC3=C2C=CC=C3Br

Specifications

Melting Point 149°C
Color White
Quantity 1 g
Formula Weight 325.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dibromodibenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.