Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dibromodibenzothiophene 98.0+%, TCI America™
SDP

Catalog No. D48221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48221G 1 g
D4822200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48221G Supplier TCI America Supplier No. D48221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 669773-34-6
Molecular Formula C12H6Br2S
Molecular Weight (g/mol) 342.05
MDL Number MFCD18451473
InChI Key ULGFJZPCGNTWFK-UHFFFAOYSA-N
Synonym 4,6-dibromodibenzo b,d thiophene, 4,6-dibromo-dibenzothiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene
PubChem CID 5245996
IUPAC Name 6,10-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
SMILES BrC1=CC=CC2=C1SC1=C2C=CC=C1Br

Specifications

Melting Point 186°C
Color White
Quantity 1 g
Formula Weight 342.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dibromodibenzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.