Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine 98.0+%, TCI America™
SDP

Catalog No. D45981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45981G 1 g
D45985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45981G Supplier TCI America Supplier No. D45981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150728-13-5
Molecular Formula C15H10Cl2N4O2
Molecular Weight (g/mol) 349.171
MDL Number MFCD03839838
InChI Key IZGOBGVYADHVKH-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-2-methoxyphenoxy-2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy-2,2-bipyrimidine, 2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxy-phenoxy-2,2' bipyrimidinyl, 2,2'-bipyrimidine,4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxyphenoxy-2-pyrimidin-2-yl pyrimidine, 4,6-dichloro-5-2-methoxyphenoxy 2,2'-bipyrimidine, 4,6-dichloro-5-o-methoxyphenoxy-2-2-pyrimidinyl pyrimidine, pubchem17752
PubChem CID 10545706
IUPAC Name 4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
SMILES COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl

Specifications

Melting Point 159°C
Color Yellow
Quantity 1 g
Formula Weight 349.17
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2′-bipyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.